EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
High-resolution NMR spectroscopy in solids by truly magic-angle spinning.
PMID 15818628
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
151114
Cholesteryl acetate, 97%
C29H48O2