EMAIL THIS PAGE TO A FRIEND
Cell
Molecular cloning of cDNAs encoding the GAP-associated protein p190: implications for a signaling pathway from ras to the nucleus.
PMID 1581965
Abstract
In mitogenically stimulated and tyrosine kinase-transformed cells, a substantial fraction of the ras GTPase-activating protein (GAP) forms a complex with a protein termed p190. We have cloned several cDNAs encoding the p190 protein. Analysis of the predicted protein sequence reveals three distinct domains with homology to previously described sequences. An N-terminal domain of p190 contains sequence motifs that are found in all of the known GTPases. At the C-terminus of the protein is a domain that contains sequences very similar to those found in the breakpoint cluster region gene product, n-chimerin, and rho GAP, all of which have been shown to possess intrinsic GAP activity on small GTPases. Finally, a 778 aa segment in the middle of p190 is nearly identical in sequence to a recently described transcriptional repressor. This raises the possibility that p190, acting via GAP, can transduce signals from p21ras to the nucleus, perhaps affecting expression of specific cellular genes.