EMAIL THIS PAGE TO A FRIEND
Journal of molecular endocrinology
Tetracycline-regulated secretion of human (pro)insulin following plasmid-mediated transfection of human muscle.
PMID 15821105
Abstract
Long-term secretion of insulin by host muscle following transduction with an insulin gene construct offers the potential of gene therapy for diabetes without immunosuppression. Clinical implementation will be dependent on proof of principle in human tissue and a system for safely regulating basal insulin levels. Liposomal co-transfection with a tetracycline-responsive wild type human preproinsulin (pTRE-hppI1) or mutant construct (pTRE-hppI4), in which PC2 and PC3 cleavage sites were altered to form tetrabasic consensus sites for furin, together with pTet-off (coding for a transactivating protein) was evaluated in the C2C12 mouse myoblast cell line and human myoblasts following establishment in primary culture. In the absence of tetracycline, (pro)insulin secretion in C2C12 and human myoblasts transfected with tetracycline-responsive hppI1 and hppI4 constructs was comparable to that following transfection with equivalent constructs under the control of a constitutively active cytomegaloviral promoter. Percentage processing to mature insulin was <5% in C2C12 and human myoblasts transfected with pTet-off/pTRE-hppI1 but >90% in C2C12 cells and 45-60% in human myoblasts on transfection with pTet-off/pTRE-hppI4. Incremental dose-responsive suppression of proinsulin secretion was demonstrated in C2C12 and human myoblasts expressing pTet-off/pTRE-hppI1 following incubation with tetracycline (0-100 microg/ml) for up to 72 h. Reversibility was confirmed following tetracycline withdrawal. Dose-responsive tetracycline-inducible repression of mature insulin secretion was confirmed in C2C12 cells following transfection with pTet-off/pTRE-hppI4. Regulation of human proinsulin biosynthesis and secretion has been attained in vivo following plasmid-mediated gene transfer to rat skeletal muscle and oral tetracycline administration. In conclusion, processing to mature insulin has been confirmed following plasmid-mediated gene transfer to human muscle in addition to in vitro- and in vivo-regulated human proinsulin secretion employing the safe and well-tolerated antibiotic, tetracycline.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87128
Tetracycline, 98.0-102.0% (HPLC)
C22H24N2O8 · xH2O