EMAIL THIS PAGE TO A FRIEND
Gut
Role of cyclooxygenases 1 and 2 in the modulation of neuromuscular functions in the distal colon of humans and mice.
PMID 15831902
Abstract
Cyclooxygenase isoforms (COX-1, COX-2) may exert differential regulatory actions on enteric motor functions under normal or pathological conditions. To examine the occurrence and functions of COX-1 and COX-2 in the neuromuscular compartment of normal distal colon using human and murine tissue. Gene expression (human, mouse), protein expression (human), gene deletion (mouse), and the effects of dual and isoform specific COX inhibitors on in vitro motility (human, mouse) were investigated. Reverse transcription-polymerase chain reaction (RT-PCR) showed mRNA expression of COX-1 and COX-2 in human and wild-type mouse colonic muscle whereas only COX-2 or COX-1 was detected in COX-1 or COX-2 knockout animals. Immunohistochemistry localised both isoforms in neurones of myenteric ganglia, COX-1 in circular layer myocytes, and COX-2 in longitudinal muscle. Indomethacin (COX-1/COX-2 inhibitor), SC-560 (COX-1 inhibitor), or DFU (COX-2 inhibitor) enhanced atropine sensitive electrically induced contractions of human longitudinal muscle. The most prominent actions were recorded with indomethacin or SC-560 plus DFU. These results were confirmed under pharmacological blockade of non-cholinergic nerves. Atropine sensitive contractions evoked by carbachol in the presence of tetrodotoxin were enhanced by indomethacin or DFU but not by SC-560. In wild-type mice, contractile responses to electrical stimulation were enhanced by indomethacin, SC-560, or DFU. SC-560 potentiated electrically induced contractions in COX-2, but not COX-1, knockout mice. In contrast, DFU enhanced the contractions elicited by electrical stimuli in COX-1, but not in COX-2, knockout mice. These results indicate that COX-1 and COX-2 are expressed in the neuromuscular compartment of normal human colon where they modulate cholinergic excitatory control of colonic motility at prejunctional and postjunctional sites, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217794
DL-3-Aminoisobutyric acid, 98%
C4H9NO2