EMAIL THIS PAGE TO A FRIEND
Journal of the American Mosquito Control Association
Efficacy of permethrin- and cyphenothrin-impregnated nettings against Culex pipiens pallens.
PMID 1583496
Abstract
Laboratory tests were conducted to determine the effect of different bed net materials, impregnated with permethrin or cyphenothrin, on Culex mosquitoes. Polyethylene, polyester and vinylon nettings were more efficacious than cotton or nylon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46037
Cyphenothrin, PESTANAL®, analytical standard
C24H25NO3