EMAIL THIS PAGE TO A FRIEND
Mikrobiologiia
[The synthesis of hydroxybutyrate and hydroxyvalerate copolymers by the bacterium Ralstonia eutropha].
PMID 15835780
Abstract
The paper deals with the study of the synthesis of 3-hydroxybutyrate (3HB) and 3-hydroxyvalerate (3HV) copolymers by the bacterium Ralstonia eutropha B-5786 grown under different carbon nutrition conditions (growth on carbon dioxide, fructose, and CO2-valerate and fructose-valerate mixtures). The parameters to be analyzed included the yield of biomass, the yield, synthesis rate, and composition of copolymers, the activity of the key enzymes of polyhydroxyalkanoate (PHA) synthesis (beta-ketothiolase, acetoacetyl-CoA reductase, and PHA synthase), the maximum tolerable concentration of valerate to the bacterium, and the conditions that govern the incorporation of hydroxyvalerate to copolymers. This allowed the relationship between cultivation conditions and the proportion of monomers in the copolymers to be deduced. We were able to synthesize a range of 3HB/3HV copolymers and found that the thermal characteristics and the degree of crystallinity of these copolymers depend on the molar fraction of 3HV.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1625
Acetoacetyl coenzyme A sodium salt hydrate, cofactor for acyl transfer
C25H40N7O18P3S · xNa+ · yH2O