EMAIL THIS PAGE TO A FRIEND
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association
Erythropoietin receptors: their role beyond erythropoiesis.
PMID 15840686