EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Two flavonoids from Artemisia herba-alba Asso with in vitro GABAA-benzodiazepine receptor activity.
PMID 15848034
Abstract
An ethyl acetate extract of Artemisia herba-alba was partitioned by HPLC in 10 fractions that were tested in the [(3)H]-flumazenil radioligand assay, for affinity to the GABA(A)-benzodiazepine receptor. Two fractions showed activity from which hispidulin and cirsilineol were isolated. The structures were confirmed by (1)H NMR. The IC(50) values were 8 microM for hispidulin and 100 microM for cirsilineol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67403
Cirsilineol, analytical standard
C18H16O7
SML0582
Hispidulin, ≥98% (HPLC)
C16H12 O6