EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Syntheses of novel heterocycles as anticancer agents.
PMID 15848764
Abstract
Several pteridine analogues 4-13, 23-26 have been synthesized and tested in vitro against three cancer cell lines, MCF7 (breast), NCI-H460 (lung) and SF-268 (CNS). All tested pteridines can serve as novel templates for the anticancer chemotherapy and can serve as new leads in cancer chemotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183946
6-(Chloromethyl)uracil, 98%
C5H5ClN2O2