EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
The O-polysaccharide from the lipopolysaccharide of Providencia stuartii O44 contains L-quinovose, a 6-deoxy sugar rarely occurring in bacterial polysaccharides.
PMID 15854616
Abstract
The O-polysaccharide (O-antigen) of Providencia stuartii O44:H4 (strain 3768/51) was obtained by mild acid degradation of the lipopolysaccharide and studied by sugar and methylation analyses along with (1)H and (13)C NMR spectroscopy, including 2D (1)H,(1)H COSY, TOCSY, ROESY, and H-detected (1)H,(13)C HSQC, and HMQC-TOCSY experiments. The O-polysaccharide was found to have a branched hexasaccharide repeating unit of the following structure: [Formula: see text].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9761
6-Deoxy-D-glucose
C6H12O5