EMAIL THIS PAGE TO A FRIEND
The Veterinary clinics of North America. Small animal practice
Advantages and guidelines for using isoflurane.
PMID 1585568
Abstract
Isoflurane offers many advantages over other inhalational anesthetics. Its faster induction and recovery, relative sparing effect on cardiovascular function and cerebral blood flow autoregulation, and negligible metabolism make this drug particularly useful in the anesthetic management of the debilitated, aged, or unusual veterinary patient.