EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Selective synthesis of C-arylglycosides via Cp*RuCl-catalyzed partially intramolecular cyclotrimerizations of C-alkynylglycosides.
PMID 15858662
Abstract
In the presence of catalytic amounts of Cp*RuCl(cod), the partially intramolecular cyclotrimerizations of various C-alkynylglycosides and C-diynylglycosides proceeded at ambient temperature to afford C-arylglycosides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208523
Ruthenium(III) chloride, Ru content 45-55%
Cl3Ru