EMAIL THIS PAGE TO A FRIEND
Toxicology
Use of the local lymph node assay in assessment of immune function.
PMID 15863253
Abstract
The murine local lymph node assay (LLNA) was originally developed as a predictive test method for the identification of chemicals with sensitizing potential. In this study we demonstrated that an adapted LLNA can also be used as an immune function assay by studying the effects of orally administered immunomodulating compounds on the T-cell-dependent immune response induced by the contact sensitizer 2,4-dinitrochlorobenzene (DNCB). C57Bl/6 mice were treated with the immunotoxic compounds cyclosporin A (CsA), bis(tri-n-butyltin)oxide (TBTO) or benzo[a]pyrene, (B[a]P). Subsequently, cell proliferation and interferon-gamma (IFN-gamma) and interleukin (IL)-4 release were determined in the auricular lymph nodes (LNs) after DNCB application on both ears. Immunosuppression induced by CsA, TBTO and B[a]P was clearly detectable in this application of the LLNA. Cytokine release measurements proved valuable to confirm the results of the cell proliferation assay and to obtain an indication of the effect on Th1/Th2 balance. We believe to have demonstrated the applicability of an adapted LLNA as an immune function assay in the mouse.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B53383
Bis(tributyltin) oxide, 96%
C24H54OSn2