EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Redefining the structure-activity relationships of 2,6-methano-3-benzazocines. Part 3: 8-Thiocarboxamido and 8-thioformamido derivatives of cyclazocine.
PMID 15863314
Abstract
8-Position variants of cyclazocine have been made where the phenolic 8-OH was replaced by thioamide, amidine, guanidine, urea and thiourea groups. High affinity for opioid receptors was observed for the 8-CSNH2 and 8-NHCHS analogues indicating that these groups are isosteric with not only the 8-OH but with the previously synthesized 8-CONH2 and 8-NHCHO cyclazocine derivatives.