EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and relative bioavailability of meptazinol benzoyl esters as prodrugs.
PMID 15863326
Abstract
Three meptazinol benzoyl esters (1-3) were synthesized as prodrugs to minimize the first-pass effect of meptazinol and improve the bioavailability. Among these three esters, compound 3 showed better bioavailability than the parent meptazinol. Further, the relative regional bioavailability of prodrug 3 was evaluated using in situ closed loop study in rats, which showed that prodrug 3 has higher absorption efficacy in rat intestine. Thusly, prodrug 3 may be worth for further development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2824
Meptazinol hydrochloride
C15H23NO· HCl