EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis and structure-activity relationships of 6-O-arylpropargyl diazalides with potent activity against multidrug-resistant Streptococcus pneumoniae.
PMID 15863336
Abstract
A novel series of 6-O-arylpropargyl diazalides was synthesized and evaluated for their antibacterial activity. Members of this series exhibited potent activity against erythromycin-resistant respiratory tract pathogens.