EMAIL THIS PAGE TO A FRIEND
Nihon Ishinkin Gakkai zasshi = Japanese journal of medical mycology
Evaluation of a newly developed identification kit, RID Zyme CAS test, for Candida albicans.
PMID 15864259
Abstract
To evaluate a newly developed identification kit, the RID Zyme CAS test for Candida albicans, 1136 C. albicans and 403 non-albicans Candida strains were tested. Distinction of medically important non-albicans strains, with the exception of C. dubliniensis, was obtained. These results show that this new kit is simple and effective for the identification of C. albicans in clinical samples. Furthermore, the one hour period for identification makes it very attractive.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9659
4-Methylumbelliferyl N-acetyl-β-D-galactosaminide, ≥98% (HPLC)
C18H21NO8