EMAIL THIS PAGE TO A FRIEND
Mutation research
Prenatal and perinatal fenarimol-induced genotoxicity in leukocytes of in vivo treated rats.
PMID 15866470
Abstract
The identification of environmental compounds that have adverse effects on reproductive health and animal development is particularly challenging. Fenarimol, a systemic fungicide, is considered non or weakly genotoxic. However, its available toxicological data are controversial and incomplete. This study was conducted in rat in vivo to determine whether this compound (150 and 300 mg/kg) had adverse effects on DNA integrity in dams and pups after maternal subcutaneous exposure. The animals were exposed during early gestation (1-6 days), late gestation (last 6 days), or first 6 days of lactation. Findings on fenarimol genotoxicity showed an adverse effect when detected by the Comet assay, both in dams and pup, and state that animal sensitivity to fenarimol is higher during postnatal period. Since the DNA damage increases during the time of exposure (2 h to 6 days after the birth), our data on pups suggest that fenarimol can mainly act on cell DNA through direct exposure of litter via milk.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45484
Fenarimol, PESTANAL®, analytical standard
C17H12Cl2N2O