EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
PROPHIS: parabolic trough-facility for organic photochemical syntheses in sunlight.
PMID 15875073
Abstract
The PROPHIS facility is an efficient tool for the synthesis of chemicals with moderately concentrated sunlight on a semi-technical scale. The feasibility of selected solar photochemical reaction classes--including heterogeneous and homogeneous reactions--has been demonstrated using various set-ups of the plant. This paper outlines the potential of solar photochemistry by representative examples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P33402
2-Phenylpyridine, 98%
C11H9N