EMAIL THIS PAGE TO A FRIEND
Water research
Dichloroaniline retention by nanofiltration membranes.
PMID 15878032
Abstract
This study evaluates the performance of two nanofiltration membranes in removing a herbicide: dichloroaniline. The membranes, one polyamide and one cellulose acetate, have a cut-off in the range 150-300 g/mol (manufacturers' data). The experiments were carried out with solutions of dichloroaniline in demineralized water, with concentrations from 1 to 10 ppb. For each membrane, the amount of herbicide retained and adsorbed by the membrane was determined as a function of feed concentration and transmembrane pressure. The two membranes, made of different materials but having the same nominal cut-off, retained dichloroaniline to very different extents and by different mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35829
2,4-Dichloroaniline, PESTANAL®, analytical standard
C6H5Cl2N
112151
2,4-Dichloroaniline, 99%
C6H5Cl2N