EMAIL THIS PAGE TO A FRIEND
Journal of the American Society for Mass Spectrometry
The structure of gas-phase bradykinin fragment 1-5 (RPPGF) ions: an ion mobility spectrometry and H/D exchange ion-molecule reaction chemistry study.
PMID 15878286
Abstract
Ion mobility-mass spectrometry (IM-MS) data is interpreted as evidence that gas-phase bradykinin fragment 1-5 (BK1-5, RPPGF) [M + H](+) ions exist as three distinct structural forms, and the relative abundances of the structural forms depend on the solvent used to prepare the matrix-assisted laser desorption ionization (MALDI) samples. Samples prepared from organic rich solvents (90% methanol/10% water) yield ions having an ion mobility arrival-time distribution (ATD) that is dominated by a single peak; conversely, samples prepared using mostly aqueous solvents (10% methanol/90% water) yield an ATD composed of three distinct peaks. The BK1-5 [M + H](+) ions were also studied by gas-phase hydrogen/deuterium (H/D) exchange ion-molecule reactions and this data supports our interpretation of the IM-MS data. Plausible structures for BK1-5 ions were generated by molecular dynamics (MD). Candidate MD-generated structures correlated to measured cross-sections suggest a compact conformer containing a beta-turn whereas a more extended, open form does not contain such an interaction. This study illustrates the importance of intra-molecular interactions in the stabilization of the gas-phase ions, and these results clearly illustrate that solution-phase parameters (i.e., MALDI sample preparation) greatly influence the structures of gas-phase ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1401
Bradykinin Fragment 1-5, ≥97% (HPLC)
C27H40N8O6