EMAIL THIS PAGE TO A FRIEND
Journal of the Indian Medical Association
Surgical manifestations of ascariasis in childhood.
PMID 1588117
Abstract
Amongst 50 patients with surgical complications of Ascaris lumbricoides 35 patients (70%) were treated conservatively and 15 patients (30%) required surgical intervention which included squeezing of worms to distal colon, resection and anastomosis of ileum, appendicectomy and peritoneal lavage. Intestinal perforation was observed in 3 cases. Two patients (4%) died because of postoperative complications. Those presenting with subacute intestinal obstruction had no mortality or morbidity. Although a large number of roundworms were seen in the jejunum, the obstruction was generally observed in the distal ileum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46632
Citric acid sesquipiperazine salt hydrate, VETRANAL®, analytical standard
C6H8O7 · 1.5C4H10N2 · xH2O