EMAIL THIS PAGE TO A FRIEND
JAMA
Effects of a 5-lipoxygenase-activating protein inhibitor on biomarkers associated with risk of myocardial infarction: a randomized trial.
PMID 15886380
Abstract
Myocardial infarction (MI) is the leading cause of death in the world. Variants in the 5-lipoxygenase-activating protein (FLAP) gene are associated with risk of MI. To determine the effect of an inhibitor of FLAP on levels of biomarkers associated with MI risk. A randomized, prospective, placebo-controlled, crossover trial of an inhibitor of FLAP (DG-031) in MI patients who carry at-risk variants in the FLAP gene or in the leukotriene A4 hydrolase gene. Of 268 patients screened, 191 were carriers of at-risk variants in FLAP (87%) or leukotriene A4 hydrolase (13%). Individuals were enrolled in April 2004 and were followed up by designated cardiologists from a university hospital in Iceland until September 2004. Patients were first randomized to receive 250 mg/d of DG-031, 500 mg/d of DG-031, 750 mg/d of DG-031, or placebo. After a 2-week washout period, patients received DG-031 if they had received placebo first or placebo if they had received DG-031 first. Treatment periods lasted for 4 weeks. Changes in levels of biomarkers associated with risk of MI. In response to 750 mg/d of DG-031, production of leukotriene B4 was significantly reduced by 26% (95% confidence interval [CI], 10%-39%; P = .003) and myeloperoxidase was significantly reduced by 12% (95% CI, 2%-21%; P = .02). The higher 2 doses of DG-031 produced a nonsignificant reduction in C-reactive protein (16%; 95% CI, -2% to 31%; P = .07) at 2 weeks. However, there was a more pronounced reduction (25%; 95% CI, 5%-40%; P = .02) in C-reactive protein at the end of the washout period that persisted for another 4 weeks thereafter. The FLAP inhibitor DG-031 was well tolerated and was not associated with any serious adverse events. In patients with specific at-risk variants of 2 genes in the leukotriene pathway, DG-031 led to significant and dose-dependent suppression of biomarkers that are associated with increased risk of MI events.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0411
BAY-X-1005, ≥98% (HPLC)
C23H23NO3