EMAIL THIS PAGE TO A FRIEND
Angiology
Aminophylline: exploring cardiovascular benefits versus medical malcontent.
PMID 15889197
Abstract
Although aminophylline/theophylline has been relegated to third- and fourth-line status in the cardiopulmonary armamentarium, its use in specific pathophysiologic states, especially those of cardiac etiology, can be of significant benefit. The consulting clinician should maintain an awareness of its potential as adjunctive therapy in cases of atrioventricular block, cardiac arrest, heart failure, and bradyarrhythmias in particular. It should not yet be shelved as an archaic agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1755
Aminophylline, ≥98%, powder
C7H8N4O2 · 0.5C2H8N2