EMAIL THIS PAGE TO A FRIEND
Journal of enzyme inhibition and medicinal chemistry
Suicide inhibition of acetohydroxyacid synthase by hydroxypyruvate.
PMID 15895677
Abstract
Acetohydroxyacid synthase (Ec 2.2.1.6) catalyses the thiamine diphosphate-dependent reaction between two molecules of pyruvate yielding 2-acetolactacte and CO2. The enzyme will also utilise hydroxypyruvate with a k(cat) value that is 12% of that observed with pyruvate. When hydroxypyruvate is the substrate, the enzyme undergoes progressive inactivation with kinetics that are characteristic of suicide inhibition. It is proposed that the dihydroxyethyl-thiamine diphosphate intermediate can expel a hydroxide ion forming an enol that rearranges to a bound acetyl group.