EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Detection of sensitivity to cardol in north-eastern and south-eastern Brazil.
PMID 15899012
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91822
5-Pentadecylresorcinol, analytical standard
C21H36O2