EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Polymerase chain reaction for identification of aldoxime dehydratase in aldoxime- or nitrile-degrading microorganisms.
PMID 15899412
Abstract
We developed a molecular screening procedure using Southern hybridization and polymerase chain reaction (PCR) to identify aldoxime dehydratase (Oxd) encoding genes (oxds) among 14 aldoxime- or nitrile-degrading microorganisms. When an oxd gene of Rhodococcus erythropolis N-771 was used as a probe, positive hybridization signals were seen with the chromosomal DNA of eight strains, suggesting that these strains have similar oxd genes to R. erythoropolis N-771. By analyzing the PCR-amplified fragments with degenerate consensus primers, the occurrence of homologous Oxd coexisting with Fe-containing NHase in the active eight strains was demonstrated coinciding with the results of Southern hybridization. Whole length of oxd gene was cloned as an example from one of the positive strains, Pseudomonas sp. K-9, sequenced, and expressed in E. coli. Analysis of the primary structure of the protein (OxdK) encoded by the oxd gene of Pseudomonas sp. K-9 led to identify an Oxd having a new primary structure. Thus, the PCR-based analysis of oxd gene is a useful tool to detect and analyze the "aldoxime-nitrile pathway" in nature, since Oxd is the key enzyme for the pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1002
Acetaldehyde oxime, mixture of syn and anti, 99%
C2H5NO