EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
The rhodium catalyzed three-component reaction of diazoacetates, titanium(IV) alkoxides and aldehydes.
PMID 15900348
Abstract
The rhodium(II)-catalyzed three-component reaction of diazoacetates, titanium alkoxides and aldehydes is shown to give alpha-alkoxyl-beta-hydroxyl acid derivatives; the novel C-C bond formation reaction is proposed to occur through oxonium ylides derived from diazo compounds and titanium alkoxides, and followed by intermolecular trapping by aldehydes.