EMAIL THIS PAGE TO A FRIEND
The Journal of Comparative Neurology
Chain formation and glial tube assembly in the shift from neonatal to adult subventricular zone of the rodent forebrain.
PMID 15906315
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse