EMAIL THIS PAGE TO A FRIEND
Life sciences
Nitric oxide involvement in the delayed antiarrhythmic effect of treadmill exercise in dogs.
PMID 15907332
Abstract
We have shown previously that a single period of treadmill exercise in dogs protects the heart against the severe ventricular arrhythmias that arise when a major (anterior descending) branch of the left coronary artery is occluded following anaesthesia 24 h later. This protection is aminoguanidine sensitive, suggesting a role for nitric oxide (NO) in this exercise-induced delayed antiarrhythmic effect. The present study has further examined the possible role of NO as a mediator and/or as a trigger using the selective induced (iNOS) inhibitor S-(2-aminoethyl)-methyl-isothiourea (AEST) and the specific but not selective nitric oxide synthase inhibitor N(omega)-nitro-L-arginine-methyl-ester (L-NAME). Exercise markedly reduced the severity of ischaemia and reperfusion-induced ventricular arrhythmias 24 h later. Thus, only one of the dogs (8%) so exercised fibrillated on occlusion (contrast 46% in the control, non-exercised dogs; P<0.05) and the marked changes in the inhomogeneity of electrical activation that occur in the ischaemic region following occlusion were much reduced (P<0.05 compared to controls). This delayed exercise-induced cardioprotection was significantly attenuated by the nitric oxide synthase (NOS) inhibitors L-NAME, given prior to the exercise protocol and by AEST given prior to the coronary artery occlusion. For example, survival from the ischaemia-reperfusion insult was 54% in the exercise dogs, 0% in the controls and 14% in those dogs given a NOS inhibitor. We conclude that nitric oxide (NO) is both the trigger and the mediator of this delayed protection against ischaemia and reperfusion-induced arrhythmias.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5879
2-(2-Aminoethyl)isothiourea dihydrobromide
C3H9N3S · 2HBr
A54601
2-(2-Aminoethyl)isothiourea dihydrobromide, 99%
C3H9N3S · 2HBr