EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Simultaneous rapid quantification of ginsenoside Rg1 and its secondary glycoside Rh1 and aglycone protopanaxatriol in rat plasma by liquid chromatography-mass spectrometry after solid-phase extraction.
PMID 15907630
Abstract
Ginseng saponins isolated from ginseng, have been regarded as the principal constituents responsible for the biological activities. The aim of this study was to set up a liquid chromatography-mass spectrometry (LC-MS) method for simultaneously determine the concentration of Ginsenoside Rg1 and its secondary glycoside Rh1 and aglycone protopanaxatriol (PPT) in rat plasma so as to study the pharmacokinetics of Rg(1) after intraveneous (i.v.) and intragastric gavage (i.g.) administration. One hundred microliters or 1.0 ml of rat plasma samples from i.v. or i.g. treated rats were used respectively for analysis. After solid-phase extraction (SPE) and high performance liquid chromatography (HPLC) separation, the chloride adduct anions [M+Cl]- of Rg1, Rh1 and PPT were analyzed by LC-MS in selected ions monitoring (SIM) mode. Rg(1) could be determined by this LC-MS method over the ranges of 1.56-250 ng/ml and 250-20,000 ng/ml with the correlation coefficients of 0.999 and 0.9998, respectively. The detection limits (LOD) of this method was 20 pg (S/N>3) for Rg1, 100 pg for Rh1 and 10 pg for PPT. Chromatographic separation was achieved in less than 8 mins. The method has been used for the pharmacokinetic study of Rg1 in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56805
Ginsenoside Rh1, analytical standard
C36H62O9