EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Immunomodulatory activity of cucurbitacin E isolated from Ecballium elaterium.
PMID 15908139
Abstract
The immunomodulatory effect of cucurbitacin E, extracted from Ecballium elaterium, was tested on peripheral human lymphocytes. These lymphocytes were co-cultured with cancer cells and an interesting lymphocyte-mediated cytotoxicity was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0577 Cucurbitacin E, ≥95% (HPLC)
C32H44O8