EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Heteroarylisopropylamines as MAO inhibitors.
PMID 15908219
Abstract
The in vitro monoamine oxidase inhibitory (MAOI) activities of 11 heteroarylisopropylamines vis-à-vis MAO-A and MAO-B were described and interpreted in terms of possible interactions with the enzyme active site. Molecular dynamics simulations allowed a comparison between the most active MAO-A inhibitor of the series, the 1-(2-benzofuryl)-2-aminopropane, and the specific, analogous MAO-A substrate serotonin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471291
Isopropylamine, ≥99.5%
C3H9N
45945
Isopropylamine, anhydrous, analytical standard
C3H9N