EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Interaction between gamma-aminobutyric acid GABAB and cannabinoid CB1 receptors in spinal pain pathways in rat.
PMID 15910802
Abstract
Antinociceptive effects of cannabinoids are mediated, in part, at the spinal level. Cannabinoid CB1 receptors are co-localized with dorsal horn interneurons containing gamma-aminobutyric acid (GABA). In this study, we investigated the interaction between intrathecally administered cannabinoid and GABA(B) receptor agonists and antagonists in the modulation of formalin-induced pain at the spinal level. Intrathecal pretreatment of rats with a cannabinoid receptor antagonist [N-(piperidin-1-yl)-5-(4-chlorophenyl)-1-(2,4-dichlorophenyl)-4-methyl-1-H-pyrazole-3-carboxamide] (SR141716A, 30 microg) decreased the analgesic effect of the intrathecal administration of the GABA(B) receptor agonist, baclofen (0.125 microg and 0.25 microg). Intrathecal administration of the GABA(B) receptor antagonist, saclofen (30 microg), 10 min before administration of the cannabinoid receptor agonist (-)-cis-3-[2-hydroxy-4-(1,1-dimethylheptyl)-phenyl]-trans-4-(3-hydroxy-propyl)-cyclohexano (CP55940), did not affect the analgesia produced by the cannabinoid receptor agonist. Our results confirm that intrathecal administration of cannabinoid and GABA(B) receptor agonists have analgesic effects and that spinal antinociceptive effects of GABA(B) receptor agonists are likely through endocannabinoid modulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S166
Saclofen, solid
C9H12ClNO3S