EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and characterization of azole isoflavone inhibitors of aromatase.
PMID 15911319
Abstract
The synthesis and biological evaluation of a series of 2-azole and 2-thioazole isoflavones as potential aromatase inhibitors are described. Differences in inhibitory activity of triazole and imidazole inhibitors are rationalized with density functional theory to expose a key difference in the electronic structure of these molecules. In addition, difference binding spectra of inhibitors to immunoaffinity-purified aromatase produces classical Type II spectra consistent with coordination of the nitrogen lone pair electrons to the aromatase P450 heme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9657
DL-Aminoglutethimide
C13H16N2O2