EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthetic iminosugar derivatives as new potential immunosuppressive agents.
PMID 15916418
Abstract
Several iminosugar derivatives were synthesized, and their effects on the secretion of IL-4 and IFN-gamma from the mouse splenocytes were examined. The effects on membrane expression of other T cell-associated molecules (CD3, CD4, CD8) and B cell-associated molecules (CD19) were also investigated. The experimental data demonstrated that synthetic iminosugars hold potential as immunosuppressive agents.