EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Hydrolyzable tannins: potent inhibitors of hydroperoxide production and tumor promotion in mouse skin treated with 12-O-tetradecanoylphorbol-13-acetate in vivo.
PMID 1592533
Abstract
The anti-oxidant and the anti-tumor-promotion activities of several hydrolyzable tannins (HTs), including a commercial tannic-acid (TA) mixture, were examined in mouse skin treated with 12-O-tetradecanoylphorbol-13-acetate (TPA) in vivo. A single application of TPA gradually increases the hydroperoxide (HPx)-producing activity of the epidermis, which is maximally stimulated at 3 days and returns to control levels at 9 days. Pre-treatments with TA and ellagic acid (EA) strongly inhibit, in a dose-dependent manner, this HPx response to TPA. Total inhibition by TA lasts for about 16 hr, beyond which it is substantially reduced but not completely lost. TA can also reduce the level of epidermal HPx when it is applied 36 hr after the tumor promoter. EA is an antioxidant 10 times more potent than TA and n-propyl gallate (PG), which are equally effective against TPA-induced HPx production. Gallic acid is the least effective of the HTs in inhibiting HPx formation. TA also inhibits the production of HPx induced by several structurally different tumor promoters and the greater HPx responses produced by repeated TPA treatments. When applied 20 min before each promotion treatment, twice a week for 45 weeks, several HTs inhibit the incidence and yield of papillomas and carcinomas promoted by TPA in initiated skin. Overall, TA is more effective than EA and PG in inhibiting skin-tumor promotion by TPA, suggesting that the anti-oxidant effects of HTs are essential but not sufficient for their anti-tumor-promotion activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2250
Ellagic acid, ≥95% (HPLC), powder, from tree bark
C14H6O8