EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Lack of protease activated receptor (PAR) expression in purified human basophils.
PMID 15928814