EMAIL THIS PAGE TO A FRIEND
MMWR. Morbidity and mortality weekly report
Human exposure to mosquito-control pesticides--Mississippi, North Carolina, and Virginia, 2002 and 2003.
PMID 15931155
Abstract
Public health officials weigh the risk for mosquito-borne diseases against the risk for human exposure to pesticides sprayed to control mosquitoes. Response to outbreaks of mosquito-borne diseases has focused on vector control through habitat reduction and application of pesticides that kill mosquito larvae. However, in certain situations, public health officials control adult mosquito populations by spraying ultra-low volume (ULV) (<3 fluid ounces per acre [oz/acre]) mosquito-control (MC) pesticides, such as naled, permethrin, and d-phenothrin. These ULV applications generate aerosols of fine droplets of pesticides that stay aloft and kill mosquitoes on contact while minimizing the risk for exposure to persons, wildlife, and the environment. This report summarizes the results of studies in Mississippi, North Carolina, and Virginia that assessed human exposure to ULV naled, permethrin, and d-phenothrin used in emergency, large-scale MC activities. The findings indicated ULV application in MC activities did not result in substantial pesticide exposure to humans; however, public health interventions should focus on the reduction of home and workplace exposure to pesticides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45429
Dibrom®, PESTANAL®, analytical standard
C4H7Br2Cl2O4P
36193
Phenothrin, PESTANAL®, analytical standard
C23H26O3