EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of isocoumarins via Pd/C-mediated reactions of o-iodobenzoic acid with terminal alkynes.
PMID 15932318
Abstract
The coupling reaction of o-iodobenzoic acid with terminal alkynes by using a catalyst system of 10% Pd/C-Et3N-CuI-PPh3 has been studied in a variety of solvents. 3-Substituted isocoumarins were formed in good yields and with good regioselectivity when the reaction was performed in EtOH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
418463
2-Iodobenzoic acid, ≥99%, purified by sublimation
C7H5IO2
I7675
2-Iodobenzoic acid, 98%
C7H5IO2