EMAIL THIS PAGE TO A FRIEND
Pharmacological research
COX-1 and COX-2 inhibition in horse blood by phenylbutazone, flunixin, carprofen and meloxicam: an in vitro analysis.
PMID 15939622
Abstract
We report on the inhibitory activity of the NSAIDs meloxicam, carprofen, phenylbutazone and flunixin, on blood cyclooxygenases in the horse using in vitro enzyme-linked assays. As expected, comparison of IC50 indicated that meloxicam and carprofen are more selective inhibitors of COX-2 than phenylbutazone and flunixin; meloxicam was the most advantageous for horses of four NSAIDs examined. However at IC80, phenylbutazone (+134.4%) and flunixin (+29.7%) had greater COX-2 selectivity than at IC50, and meloxicam (-41.2%) and carprofen (-12.9%) had lower COX-2 selectivity than at IC50. We therefore propose that the selectivity of NSAIDs should be assessed at the 80% as well as 50% inhibition level.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33586
Flunixin, VETRANAL, analytical standard
C14H11F3N2O2