EMAIL THIS PAGE TO A FRIEND
Heart failure reviews
Autonomic effects of spironolactone and MR blockers in heart failure.
PMID 15947893
Abstract
Neurohormonal dysfunction is an important and potentially modifiable part of the disease process of heart failure. In this review we will discuss the ways in which the autonomic system is deranged in congestive cardiac failure and the different ways we have of monitoring these abnormalities i.e.: heart rate variability, plasma norephinephrine activity, MIBG scanning, heart rate turbulence, baroreceptor function and briefly, microneurographic techniques and QT interval analysis. We will then discuss the direct effects of aldosterone and of aldosterone blockade on some of these parameters. We conclude that neurohormonal dysfunction is an important component of chronic heart failure, which is affected by aldosterone and can be modified by the use of aldosterone receptor blockade.
Related Materials