EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Identification of metalliferous ecotypes of Cistus ladanifer L. using RAPD markers.
PMID 15948588
Abstract
The genetic diversity of Cistus ladanifer ssp. ladanifer (Cistaceae) growing on ultramafic and non-ultramafic (basic and schists) soils in the NE of Portugal was studied in order to identify molecular markers that could distinguish the metal-tolerant ecotypes of this species. Random Amplified Polymorphic DNA (RAPD) markers were used in order to estimate genetic variation and differences between populations. The RAPD dataset was analysed by means of a cluster analysis and an analysis of molecular variance (AMOVA). Our results indicate a significant partitioning of molecular variance between ultramafic and non-ultramafic populations of Cistus ladanifer, although the highest percentage of this variance was found at the intra-population level. Mantel's test showed no relationship between inter-population genetic and geographic distances. A series of RAPD bands that could be related to heavy metal tolerance were observed. The identification of such markers will enable the use of Cistus ladanifer in phytoremediation procedures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11855638001 DNA Molecular Weight Marker XVI (250 bp ladder)