EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of poly-O-sulfated glycosides of 2,5-anhydro-D-mannitol.
PMID 15953593
Abstract
2,5-Anhydro-3-O-beta-D-glucopyranosyl-; -3-O-alpha-L-idopyranosyl-; -3-O-alpha-D-arabinopyranosyl-; -3-O-alpha-L-arabinopyranosyl-; -3-O-beta-D-maltopyranosyl-; -3-O-beta-D-gentiobiopyranosyl-; -1,6-di-O-beta-D-glucopyranosyl-; -1,6-di-O-alpha-L-idopyranosyl-; -1-O-beta-D-maltopyranosyl-; -1,3,6-tri-O-beta-D-glucopyranosyl-; -1,6-di-O-beta-maltopyranosyl- and -1,6-di-O-beta-D-gentiobiopyranosyl-2,5-anhydro-D-mannitol as well as their poly-O-sulfated derivatives were synthesized. The IP3-IC50 values of their sodium and/or potassium salts were determined for structure-activity studies aiming at the synthesis of new, orally active antiasthmatic compounds.