EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological evaluation of benzothiazole derivatives as potent antitumor agents.
PMID 15955697
Abstract
Based on 2-methyl-4-nitro-2H-pyrazole-3-carboxylic acid[2-(cyclohexanecarbonylamino)benzothiazol-6-yl]amide (1), which shows selective cytotoxicity against tumorigenic cell lines, 2,6-dichloro-N-[2-(cyclopropanecarbonylamino)benzothiazol-6-yl]benzamide (13b) was designed and synthesized as a biologically stable derivative containing no nitro group. The highly potent derivative 13b exhibited excellent in vivo inhibitory effect on tumor growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
190241
2-Amino-6-nitrobenzothiazole, ≥97%
C7H5N3O2S