EMAIL THIS PAGE TO A FRIEND
Anaesthesia and intensive care
Growth of Staphylococcus epidermidis in anaesthetic resuscitative drugs: implications for potential contamination.
PMID 15957694
Abstract
This controlled laboratory study investigated the survival of Staphylococcus epidermidis (S. epidermidis) over a 24 hour period in four commonly drawn-up anaesthetic resuscitative drugs: suxamethonium, atropine, metaraminol and ephedrine. These drugs were prepared in typical therapeutic concentrations and then inoculated with S. epidermidis. Samples of the inoculated drug preparations were cultured on horse blood agar plates at 0, 2, 6, 12 and 24 hourly intervals and incubated for 24 hours. Colony counts were performed at the end of the incubation period. Suxamethonium, atropine and metaraminol all showed an inhibitory effect on colony counts within the first six hours. There was a more gradual decline of colony counts over a 24 hour period in the ephedrine solution. This decline was similar to that occurring in the normal saline. It is concluded that suxamethonium, atropine and metaraminol do not support the survival of the common skin contaminant, S. epidermidis over a 24 hour period.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4773
Metaraminol (+)-bitartrate salt
C9H13NO2 · C4H6O6