EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of a laterally branched polyamine from alpha-methylene-gamma-butyrolactone.
PMID 15957916
Abstract
[reaction: see text] A polyamine derivative was prepared from alpha-methylene-gamma-butyrolactone. This method used Michael addition and lactone aminolysis followed by the nucleophilic substitution of the hydroxyl group by an azido group. The coupling of a lipophilic alkyne led to a polyamine that will be probed as a gene transfer agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226416
α-Methylene-γ-butyrolactone, 97%
C5H6O2