EMAIL THIS PAGE TO A FRIEND
Molecular and Cellular Proteomics
Proteomic identification of insulin-like growth factor-binding protein-6 induced by sublethal H2O2 stress from human diploid fibroblasts.
PMID 15958393