EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
The cationic cascade route to longifolene.
PMID 15960516
Abstract
W. S. Johnson's total synthesis of the sesquiterpenoid longifolene is a classic example of the power of cationic polycyclizations for constructing complex molecular architectures. Herein we revisit the key polycyclization step of this synthesis using hybrid Hartree-Fock/density functional theory calculations and validate the feasibility of Johnson's proposed mechanism. We also explore perturbations to the 3-center 2-electron bonding array in a key intermediate that result from changing the polycyclic framework in which it is embedded.