EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Gold layer-based dual crosslinking procedure of glucose oxidase with ferrocene monocarboxylic acid provides a stable biosensor.
PMID 15961058
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
106887
Ferrocenecarboxylic acid, 97%
C11H10FeO2